(3-Hydroxyphenyl)acetonitrile 95%

Supplier: Ambeed
Warning

A129135-1G A129135-5G A129135-100MG A129135-250MG
76718-888EA 45 USD
76718-888 76718-890 77878-936 77878-937
(3-Hydroxyphenyl)acetonitrile 95%
(3-Hydroxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₇NO
MDL Number: MFCD03265300
CAS Number: 25263-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR