(3S,4aS,8aS)-N-(tert-Butyl)-2-((2R,3R)-2-hydroxy-3-(3-hydroxy-2-methylbenzamido)-4-(phenylthio)butyl)decahydroisoquinoline-3-carboxamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Nelfinavir
A128893-1MG
A128893-2MG
A128893-5MG
76718-876EA
37.14
USD
76718-876
76718-878
76718-880
(3S,4aS,8aS)-N-(tert-Butyl)-2-((2R,3R)-2-hydroxy-3-(3-hydroxy-2-methylbenzamido)-4-(phenylthio)butyl)decahydroisoquinoline-3-carboxamide 98%
(3S,4aS,8aS)-N-(tert-Butyl)-2-((2R,3R)-2-hydroxy-3-(3-hydroxy-2-methylbenzamido)-4-(phenylthio)butyl)decahydroisoquinoline-3-carboxamide
Precaución: for Research Use Only
Formula:
C₃₂H₄₅N₃O₄S MW: 567.78 g/mol |
MDL Number:
MFCD01938163 CAS Number: 159989-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...