(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride 98%

Supplier: Ambeed
Warning

A128314-100G A128314-10G A128314-25G A128314-500G A128314-5G
76718-854EA 22 USD
76718-854 76718-856 76718-858 76718-860 76718-862
(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride 98%
(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₂·HCl
MDL Number: MFCD00038992
CAS Number: 14907-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR