1-Boc-4-piperidineacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: [1-(tert-Butoxycarbonyl)piperidin-4-yl]acetic acid, 2-(1-(tert-Butoxycarbonyl)piperidin-4-yl)acetic acid, 1-(tert-Butoxycarbonyl)-4-piperidylacetic acid

A127744-100G A127744-10G A127744-1G A127744-25G A127744-5G
76718-810EA 195.21 USD
76718-810 76718-812 76718-814 76718-816 76718-818
1-Boc-4-piperidineacetic acid 97%
1-Boc-4-piperidineacetic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MW: 243.3000 g/mol
MDL Number: MFCD00800239
CAS Number: 157688-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR