N(α)-Cbz-DL-histidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Z-DL-His-OH, Nα-Benzyloxycarbonyl-DL-histidine
A127722-10G
A127722-1G
A127722-25G
A127722-5G
76718-802EA
52
USD
76718-802
76718-804
76718-806
76718-808
N(α)-Cbz-DL-histidine 98%
N(α)-Cbz-DL-histidine
Precaución: for Research Use Only
Formula:
C₁₄H₁₅N₃O₄ MW: 289.3 g/mol |
MDL Number:
MFCD00037852 CAS Number: 19728-57-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...