3-((tert-Butoxycarbonyl)amino)-2-hydroxypropanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Boc-DL-IsoSer-OH

A126912-10G A126912-1G A126912-5G
76718-774EA 422.24 USD
76718-774 76718-776 76718-778
3-((tert-Butoxycarbonyl)amino)-2-hydroxypropanoic acid 97%
3-((tert-Butoxycarbonyl)amino)-2-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₈H₁₅NO₅
MW: 205.21 g/mol
MDL Number: MFCD08275906
CAS Number: 218916-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR