4-Amino-5-chloro-N-(1-(3-methoxypropyl)piperidin-4-yl)-2,3-dihydrobenzofuran-7-carboxamide 98%

Supplier: Ambeed
Danger

A126651-100MG A126651-10MG A126651-1MG A126651-50MG A126651-5MG A126651-1G A126651-250MG A126651-5G
76718-764EA 53 USD
76718-764 76718-766 76718-768 76718-770 76718-772 77666-808 77666-810 77878-924
4-Amino-5-chloro-N-(1-(3-methoxypropyl)piperidin-4-yl)-2,3-dihydrobenzofuran-7-carboxamide 98%
4-Amino-5-chloro-N-(1-(3-methoxypropyl)piperidin-4-yl)-2,3-dihydrobenzofuran-7-carboxamide

Precaución: for Research Use Only

Formula: C₁₈H₂₆ClN₃O₃
MDL Number: MFCD09837787
CAS Number: 179474-81-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR