2-Amino-5-bromo-2'-fluorobenzophenone 97%

Supplier: Ambeed
Warning

A125604-10G A125604-1G A125604-25G A125604-5G
76718-714EA 31 USD
76718-714 76718-716 77871-566 77871-567
2-Amino-5-bromo-2'-fluorobenzophenone 97%
2-Amino-5-bromo-2'-fluorobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₉BrFNO
MDL Number: MFCD00038380
CAS Number: 1479-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR