MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulfonic acid sodium salt) 99%

Supplier: Ambeed
Warning

A125474-1000G A125474-100G A125474-500G
76718-706EA 82 USD
76718-706 76718-708 76718-710 76718-712
MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulfonic acid sodium salt) 99%
MOPSO sodium salt (3-(4-Morpholino)-2-hydroxypropanesulphonic acid sodium salt)

Precaución: for Research Use Only

Formula: C₇H₁₄NNaO₅S
MDL Number: MFCD00070007
CAS Number: 79803-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR