3-Ethylbenzaldehyde 95%

Supplier: Ambeed
Warning

A125439-250MG A125439-5G A125439-1G
76718-702EA 36.08 USD
76718-702 76718-704 77592-432
3-Ethylbenzaldehyde 95%
3-Ethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O
MW: 134.18 g/mol
MDL Number: MFCD09263482
CAS Number: 34246-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR