Methyl-o-anisate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxybenzoic acid methyl ester, Methyl 2-methoxybenzoate, o-Anisic acid methyl ester, 2-Anisic acid methyl ester

A123930-100G A123930-10G A123930-25G A123930-500G
76718-656EA 38.2 USD
76718-656 76718-658 76718-660 76718-662
Methyl-o-anisate 98%
Methyl-o-anisate

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00008423
CAS Number: 606-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR