4,4'-Di-tert-butylbenzophenone 97%

Supplier: Ambeed
Warning

A123544-1G A123544-250MG A123544-5G
76718-650EA 73 USD
76718-650 76718-652 76718-654
4,4'-Di-tert-butylbenzophenone 97%
4,4'-Di-tert-butylbenzophenone

Precaución: for Research Use Only

Formula: C₂₁H₂₆O
MDL Number: MFCD01319937
CAS Number: 15796-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR