Methyl acetoacetate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-oxobutanoate

A123052-1000G A123052-100G A123052-10G A123052-25G A123052-500G
76718-638EA 23 USD
76718-638 76718-640 76718-642 76718-644 76718-646 76718-648
Methyl acetoacetate 95%
Methyl acetoacetate

Precaución: for Research Use Only

Formula: C₅H₈O₃
MDL Number: MFCD00008784
CAS Number: 105-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR