Methyl 4-hydroxyquinazoline-7-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-7-(methoxycarbonyl)quinazoline, 4-Hydroxy-7-(methoxycarbonyl)-1,3-benzodiazine

A122603-1G A122603-250MG A122603-5G
76718-614EA 59.42 USD
76718-614 76718-616 76718-618
Methyl 4-hydroxyquinazoline-7-carboxylate 98%
Methyl 4-hydroxyquinazoline-7-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₃
MW: 204.182 g/mol
MDL Number: MFCD00681283
CAS Number: 313535-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR