3-Phenylaniline hydrochloride 96%

Supplier: Ambeed
Warning

Synonyms: 3-Biphenylamine hydrochloride, 3-Aminobiphenyl hydrochloride, [1,1'-Biphenyl]-3-amine hydrochloride

A121876-1G A121876-250MG A121876-25G A121876-5G
76718-582EA 12 USD
76718-582 76718-584 76718-586 76718-588
3-Phenylaniline hydrochloride 96%
3-Phenylaniline hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClN
MW: 205.69 g/mol
MDL Number: MFCD00068358
CAS Number: 2113-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR