N-Methyl-3-nitro-2-pyridinamine 97%

Supplier: Ambeed
Warning

A121692-1G A121692-250MG A121692-25G A121692-5G
76718-568EA 20 USD
76718-568 76718-570 76718-572 76718-574
N-Methyl-3-nitro-2-pyridinamine 97%
N-Methyl-3-nitro-2-pyridinamine

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
MDL Number: MFCD00223335
CAS Number: 4093-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR