Methyl 2-bromo-4-(bromomethyl)benzoate 95%

Supplier: Ambeed
Danger

A121575-100MG A121575-10G A121575-1G A121575-250MG A121575-25G A121575-5G
76718-556EA 12.74 USD
76718-556 76718-558 76718-560 76718-562 76718-564 76718-566
Methyl 2-bromo-4-(bromomethyl)benzoate 95%
Methyl 2-bromo-4-(bromomethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₈Br₂O₂
MW: 307.967 g/mol
MDL Number: MFCD03840738
CAS Number: 128577-48-0
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR