DL-2-Methylaminosuccinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Methylamino)succinic acid, N-DL-Me-Asp-OH

A121483-1G A121483-25G A121483-100G A121483-500G A121483-5G
76718-552EA 14 USD
76718-552 76718-554 77878-894 77878-895 77878-896
DL-2-Methylaminosuccinic acid 98%
DL-2-Methylaminosuccinic acid

Precaución: for Research Use Only

Formula: C₅H₉NO₄
MW: 147.129 g/mol
MDL Number: MFCD00065918
CAS Number: 17833-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR