N1-Benzylpropane-1,3-diamine 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Benzylamino)propylamine, N-Benzyltrimethylenediamine, N-Benzyl-1,3-propanediamine

A121370-100G A121370-1G A121370-5G
76718-546EA 501 USD
76718-546 76718-548 76718-550
N1-Benzylpropane-1,3-diamine 98%
N1-Benzylpropane-1,3-diamine

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂
MDL Number: MFCD00274361
CAS Number: 13910-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR