Diethyl-2-propyl-1H-imidazole-4,5-dicarboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Propyl-1H-imidazole-4,5-dicarboxylic acid diethyl ester, Diethyl 2-propyl-1H-imidazole-4,5-dicarboxylate
A120433-100G
A120433-10G
A120433-25G
A120433-500G
A120433-5G
76718-502EA
25.47
USD
76718-502
76718-504
76718-506
76718-508
76718-510
Diethyl-2-propyl-1H-imidazole-4,5-dicarboxylate 95%
Diethyl-2-propyl-1H-imidazole-4,5-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₈N₂O₄ MW: 254.2800 g/mol |
MDL Number:
MFCD08062365 CAS Number: 144689-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...