Sodium phosphonoformate hexahydrate 98%

Supplier: Ambeed
Warning

Synonyms: Sodium phosphonatoformate hexahydrate, Foscarnet trisodium hexahydrate

A120238-10G A120238-1G A120238-5G A120238-25G A120238-100G
76718-494EA 21 USD
76718-494 76718-496 76718-498 76718-500 77666-720 77878-891
Sodium phosphonoformate hexahydrate 98%
Sodium phosphonoformate hexahydrate

Precaución: for Research Use Only

Formula: Na₂O₃PCO₂Na·6H₂O
MW: 300.08 g/mol (191.98 anhy)
MDL Number: MFCD00150176
CAS Number: 34156-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR