Pirenzepine dihydrochloride 99+%

Supplier: Ambeed
Warning

Synonyms: 11-(2-(4-Methylpiperazin-1-yl)acetyl)-5,11-dihydro-6H-benzo[e]pyrido[3,2-b][1,4]diazepin-6-one dihydrochloride

A119911-100MG A119911-1G A119911-250MG A119911-25MG A119911-5G
76718-472EA 14 USD
76718-472 76718-474 76718-476 76718-478 76718-480
Pirenzepine dihydrochloride 99+%
Pirenzepine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₉H₂₁N₅O₂·2HCl
MW: 424.32 g/mol
MDL Number: MFCD00055214
CAS Number: 29868-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR