5-Methoxy-4'-(trifluoromethyl)valerophenone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Methoxy-1-(4-(trifluoromethyl)phenyl)pentan-1-one, 4-(5-Methoxypentanoyl)benzotrifluoride, Fluvoxamine Maleate EP Impurity D, 5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentan-1-one
A119845-25G
A119845-50MG
A119845-5G
A119845-10MG
A119845-1G
A119845-250MG
76718-466EA
256.74
USD
76718-466
76718-468
76718-470
77592-334
77599-726
77666-718
5-Methoxy-4'-(trifluoromethyl)valerophenone 98%
5-Methoxy-4'-(trifluoromethyl)valerophenone
Precaución: for Research Use Only
Formula:
C₁₃H₁₅F₃O₂ MW: 260.252 g/mol |
MDL Number:
MFCD06658173 CAS Number: 61718-80-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...