2-(Bromomethyl)benzeneboronic acid pinacol ester 97%

Supplier: Ambeed
Danger

Synonyms: 2-(2-(bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-[2-(Bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(Bromomethyl)phenylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl bromide

A119114-10G A119114-1G A119114-250MG A119114-5G A119114-25G
76718-454EA 121 USD
76718-454 76718-456 76718-458 76718-460 77666-702
2-(Bromomethyl)benzeneboronic acid pinacol ester 97%
2-(Bromomethyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BBrO₂
MDL Number: MFCD01632210
CAS Number: 377780-72-8
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR