Methyl carbamate 95%

Supplier: Ambeed
Warning

Synonyms: Carbamic acid methyl ester

A118934-1000G A118934-100G A118934-25G A118934-500G
76718-446EA 52 USD
76718-446 76718-448 76718-450 76718-452
Methyl carbamate 95%
Methyl carbamate

Precaución: for Research Use Only

Formula: C₂H₅NO₂
MDL Number: MFCD00007964
CAS Number: 598-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR