N-Octyl-β-D-glucopyranoside 96%

Supplier: Ambeed
Warning

Synonyms: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside

A118901-25G A118901-5G A118901-100G A118901-1G
76718-442EA 203 USD
76718-442 76718-444 77599-712 77599-714
N-Octyl-β-D-glucopyranoside 96%
N-Octyl-β-D-glucopyranoside

Precaución: for Research Use Only

Formula: C₁₄H₂₈O₆
MW: 292.37 g/mol
Melting Pt: 110
MDL Number: MFCD00063288
CAS Number: 29836-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR