N-Butylbenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: N-Benzenesulphonylbutylamine

A118880-100G A118880-25G A118880-500G
76718-432EA 6.37 USD
76718-432 76718-434 76718-436
N-Butylbenzenesulfonamide 98%
N-Butylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂S
MW: 213.30 g/mol
MDL Number: MFCD00025024
CAS Number: 3622-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR