Ambroxol hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
trans-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol Hydrochloride, Ambroxol HCl, trans-4-((2-Amino-3,5-dibromobenzyl)amino)cyclohexanol hydrochloride
A118627-10G
A118627-1G
A118627-25G
A118627-50MG
A118627-100G
A118627-5G
A118627-500G
76718-424EA
31
USD
76718-424
76718-426
76718-428
76718-430
77571-106
77599-704
77666-700
Ambroxol hydrochloride 97%
Ambroxol hydrochloride
Precaución: for Research Use Only
Formula:
C₁₃H₁₈Br₂N₂O.HCl |
MDL Number:
MFCD00078932 CAS Number: 23828-92-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...