2-(3,4-Bis(4-methoxyphenyl)isoxazol-5-yl)acetic acid 95%

Supplier: Ambeed
Warning

A118517-10MG A118517-1MG A118517-25MG A118517-50MG A118517-5MG
76718-414EA 231.28 USD
76718-414 76718-416 76718-418 76718-420 76718-422
2-(3,4-Bis(4-methoxyphenyl)isoxazol-5-yl)acetic acid 95%
2-(3,4-Bis(4-methoxyphenyl)isoxazol-5-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₇NO₅
MW: 339.34 g/mol
MDL Number: MFCD00712149
CAS Number: 78967-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR