Butyl methanesulfonate 98% contains 4% DCM at maximum

Supplier: Ambeed
Warning

A118181-100G A118181-10G A118181-1G A118181-25G A118181-5G
76718-390EA 162.32 USD
76718-390 76718-392 76718-394 76718-396 76718-398
Butyl methanesulfonate 98% contains 4% DCM at maximum
Butyl methanesulfonate

Precaución: for Research Use Only

Formula: C₅H₁₂O₃S
MW: 152.212 g/mol
MDL Number: MFCD00041257
CAS Number: 1912-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR