2-(Benzyl(methyl)amino)ethanol 95%

Supplier: Ambeed
Warning

A117419-100G A117419-10G A117419-25G A117419-500G
76718-374EA 25.47 USD
76718-374 76718-376 76718-378 76718-380
2-(Benzyl(methyl)amino)ethanol 95%
2-(Benzyl(methyl)amino)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO
MW: 165.2322 g/mol
MDL Number: MFCD00002847
CAS Number: 101-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR