PNU-120596 99+%

Supplier: Ambeed
Warning

Synonyms: N-(5-Chloro-2,4-dimethoxyphenyl)-N’-(5-methyl-3-isoxazolyl)urea, 1-(5-Chloro-2,4-dimethoxyphenyl)-3-(5-methylisoxazol-3-yl)urea

A116736-100MG A116736-10MG A116736-1MG A116736-50MG A116736-5MG
76718-346EA 337.37 USD
76718-346 76718-348 76718-350 76718-352 76718-354
PNU-120596 99+%
PNU-120596

Precaución: for Research Use Only

Formula: C₁₃H₁₄ClN₃O₄
MW: 311.7 g/mol
MDL Number: MFCD00095313
CAS Number: 501925-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR