3-Phenoxybenzoic acid 98%

Supplier: Ambeed
Warning

A116615-100G A116615-10G A116615-1G A116615-25G A116615-5G
76718-330EA 204.76 USD
76718-330 76718-332 76718-334 76718-336 76718-338
3-Phenoxybenzoic acid 98%
3-Phenoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
MDL Number: MFCD00002498
CAS Number: 3739-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR