N-Butyldeoxynojirimycin 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2R,3R,4R,5S)-1-Butyl-2-(hydroxymethyl)piperidine-3,4,5-triol, N-(n-Butyl)-1-deoxynojirimycin, Miglustat
A116147-1MG
A116147-5MG
A116147-10MG
A116147-100MG
A116147-50MG
76718-310EA
16.98
USD
76718-310
76718-312
77599-676
77666-674
77666-676
N-Butyldeoxynojirimycin 98%
N-Butyldeoxynojirimycin
Precaución: for Research Use Only
Formula:
C₁₀H₂₁NO₄ MW: 219.28 g/mol |
MDL Number:
MFCD00272581 CAS Number: 72599-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...