6-Fluoro-5-methyl-4-pyrimidinamine 95%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-5-methylpyrimidin-4-amine

A115903-100MG A115903-1G A115903-250MG
76718-302EA 29.71 USD
76718-302 76718-304 76718-306
6-Fluoro-5-methyl-4-pyrimidinamine 95%
6-Fluoro-5-methyl-4-pyrimidinamine

Precaución: for Research Use Only

Formula: C₅H₆FN₃
MW: 127.12 g/mol
MDL Number: MFCD10000602
CAS Number: 18260-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR