N-Methyl-2-nitrobenzenesulfonamide 97%

Supplier: Ambeed
Warning

A115841-1G A115841-250MG A115841-5G A115841-10G
76718-296EA 44.56 USD
76718-296 76718-298 76718-300 77666-672
N-Methyl-2-nitrobenzenesulfonamide 97%
N-Methyl-2-nitrobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₇H₈N₂O₄S
MW: 216.2100 g/mol
MDL Number: MFCD01115621
CAS Number: 23530-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR