(S)-N6-Propyl-4,5,6,7-tetrahydrobenzo[d]thiazole-2,6-diamine dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Pramipexole dihydrochloride

A115399-10MG A115399-1G A115399-1MG A115399-250MG A115399-50MG A115399-5G
76718-280EA 8 USD
76718-280 76718-282 76718-284 76718-286 76718-288 77878-867
(S)-N6-Propyl-4,5,6,7-tetrahydrobenzo[d]thiazole-2,6-diamine dihydrochloride 98%
(S)-N6-Propyl-4,5,6,7-tetrahydrobenzo[d]thiazole-2,6-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₉Cl₂N₃S
MW: 284.25 g/mol
MDL Number: MFCD00876894
CAS Number: 104632-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR