Methyl 2-(2,3-dichlorobenzylidene)-3-oxobutanoate 95%

Supplier: Ambeed
Warning

A114994-10G A114994-25G
76718-276EA 18.04 USD
76718-276 76718-278
Methyl 2-(2,3-dichlorobenzylidene)-3-oxobutanoate 95%
Methyl 2-(2,3-dichlorobenzylidene)-3-oxobutanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₀Cl₂O₃
MW: 273.11 g/mol
MDL Number: MFCD00800930
CAS Number: 74073-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR