Platinum(II) bromide 98% Pt Assay:55.0 ± 1.3%
Supplier: Ambeed
|
Danger
|
A114989-1G
A114989-250MG
A114989-5G
76718-270EA
112
USD
76718-270
76718-272
76718-274
Platinum(II) bromide 98% Pt Assay:55.0 ± 1.3%
Platinum(II) bromide
Precaución: for Research Use Only
Formula:
Br₂Pt |
MDL Number:
MFCD00011180 CAS Number: 13455-12-4 UN: 3260 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...