N-(3-Hydroxypropyl)ethylenediamine 98%

Supplier: Ambeed
Danger

Synonyms: 2-(3-Hydroxypropylamino)ethylamine, N-(2-Aminoethyl)propanolamine, 3-((2-Aminoethyl)amino)propan-1-ol

A114879-1G A114879-5G A114879-25G
76718-256EA 22 USD
76718-256 77666-668 77878-863
N-(3-Hydroxypropyl)ethylenediamine 98%
N-(3-Hydroxypropyl)ethylenediamine

Precaución: for Research Use Only

Formula: C₅H₁₄N₂O
MW: 118.18 g/mol
MDL Number: MFCD00059848
CAS Number: 56344-32-2
UN: 2735
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR