3-(Trifluoromethyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-[3-(trifluoromethyl)phenyl]-1,3,2-dioxaborolane, 3-(Trifluoromethyl)phenylboronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(3-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)benzene

A114283-1G A114283-250MG A114283-25G A114283-5G
76718-248EA 9 USD
76718-248 76718-250 76718-252 76718-254
3-(Trifluoromethyl)phenylboronic acid pinacol ester 98%
3-(Trifluoromethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BF₃O₂
MDL Number: MFCD05863925
CAS Number: 325142-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR