4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-iodoquinazolin-4-amine
A114247-1G
A114247-250MG
A114247-25G
A114247-5G
76718-244EA
6
USD
76718-244
76718-246
77570-780
77570-782
4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline 98%
4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline
Precaución: for Research Use Only
Formula:
C₂₁H₁₄ClFIN₃O |
MDL Number:
MFCD09998827 CAS Number: 231278-20-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...