Fmoc-β-(2-thienyl)-D-alanine 96%

Supplier: Ambeed
Warning

Synonyms: (R)-N-Fmoc-2-thienylalanine, Fmoc-D-Thi-OH, Fmoc-D-3-(2-Thienyl)-Ala-OH, Fmoc-3-D-Ala(2-thienyl)-OH, Fmoc-β-(2-thienyl)-D-Ala-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(thiophen-2-yl)propanoic acid, Fmoc-D-2-thienylalanine

A113955-10G A113955-1G A113955-250MG A113955-5G
76718-232EA 111 USD
76718-232 76718-234 76718-236 76718-238
Fmoc-β-(2-thienyl)-D-alanine 96%
Fmoc-β-(2-thienyl)-D-alanine

Precaución: for Research Use Only

Formula: C₂₂H₁₉NO₄S
MDL Number: MFCD00065677
CAS Number: 201532-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR