1-Pentyl-4-ethynylbenzene 97%

Supplier: Ambeed
Warning

Synonyms: 4-N-Pentylphenylacetylene, 4-n-Pentylphenylacetylene, (4-Pentylphenyl)acetylene, 1-Ethynyl-4-pentylbenzene, (4-Pentylphenyl)ethyne

A113898-100G A113898-10G A113898-1G A113898-25G A113898-5G
76718-222EA 205.82 USD
76718-222 76718-224 76718-226 76718-228 76718-230
1-Pentyl-4-ethynylbenzene 97%
1-Pentyl-4-ethynylbenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₆
MW: 172.27 g/mol
MDL Number: MFCD00173884
CAS Number: 79887-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR