N,N'-Dimethyl-1,6-diaminohexane 98%

Supplier: Ambeed
Warning

A113744-10G A113744-1G A113744-25G A113744-5G A113744-100G
76718-214EA 99 USD
76718-214 76718-216 76718-218 76718-220 77666-650
N,N'-Dimethyl-1,6-diaminohexane 98%
N,N'-Dimethyl-1,6-diaminohexane

Precaución: for Research Use Only

Formula: C₈H₂₀N₂
MDL Number: MFCD00008293
CAS Number: 13093-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR