(±)-1-(1-Naphthyl)ethylamine 98%

Supplier: Ambeed
Warning

A113145-100G A113145-10G A113145-25G A113145-5G
76718-192EA 91 USD
76718-192 76718-194 76718-196 76718-198
(±)-1-(1-Naphthyl)ethylamine 98%
(±)-1-(1-Naphthyl)ethylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₃N
MDL Number: MFCD00004014
CAS Number: 42882-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR