1-Benzyl-D-proline 97%

Supplier: Ambeed
Warning

Synonyms: (R)-1-Benzylpyrrolidine-2-carboxylic acid, N-D-Bzl-Pro-OH

A112811-1G A112811-25G A112811-5G A112811-10G
76718-170EA 18.04 USD
76718-170 76718-172 76718-174 77666-640
1-Benzyl-D-proline 97%
1-Benzyl-D-proline

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₂
MW: 205.253 g/mol
MDL Number: MFCD06656733
CAS Number: 56080-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR