Methyl-3-bromo-2-oxo-2H-pyran-5-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-bromo-2-oxo-2H-pyran-5-carboxylate
A112594-100MG
A112594-1G
A112594-250MG
A112594-5G
A112594-10G
76718-158EA
12.74
USD
76718-158
76718-160
76718-162
76718-164
77592-200
Methyl-3-bromo-2-oxo-2H-pyran-5-carboxylate 95%
Methyl-3-bromo-2-oxo-2H-pyran-5-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₅BrO₄ MW: 233.02 g/mol |
MDL Number:
MFCD00463742 CAS Number: 42933-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...