Sodium phenoxyacetate 98%

Supplier: Ambeed
Warning

Synonyms: Phenoxyacetic Acid Sodium Salt

A111977-25G A111977-100G
76718-124EA 10.61 USD
76718-124 77549-174
Sodium phenoxyacetate 98%
Sodium phenoxyacetate

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 174.13 g/mol
MDL Number: MFCD00064234
CAS Number: 3598-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR