N-Acetyltyramine 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Hydroxyphenethyl)acetamide

A111970-100MG A111970-10G A111970-5G A111970-250MG
76718-118EA 6.37 USD
76718-118 76718-120 76718-122 77666-634
N-Acetyltyramine 98%
N-Acetyltyramine

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.216 g/mol
MDL Number: MFCD01670887
CAS Number: 1202-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR